missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diaminobenzoic acid, 95%

Product Code. p-7077681 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15545976

Brand: Thermo Scientific Acros 457490010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 603-81-6 | C7H8N2O2 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-81-6
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00137818
InChI Key KKTUQAYCCLMNOA-UHFFFAOYSA-N
Synonym diaminobenzoic acid, benzoic acid, diamino, 3-carboxy-1,2-diaminobenzene, pubchem3145, acmc-209mjn, 2,3-diaminobenzonic acid, 2,3-diamino benzoic acid, ksc203g4l, pharmabridge p-2952, 2,3-diaminobenzoic acid
PubChem CID 198069
IUPAC Name 2,3-diaminobenzoic acid
SMILES NC1=CC=CC(C(O)=O)=C1N

Specifications

Melting Point 232.0°C to 234.5°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 152.15
Percent Purity 95%
Chemical Name or Material 2,3-Diaminobenzoic acid
compliance-icons
Product Identifier
  • 2,3-Diaminobenzoic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.