missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-1,4-naphthoquinone, 98%

Product Code. 11475363
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475363 500 g 500g
11474110 25 g 25g
11465363 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475363 Supplier Thermo Scientific Alfa Aesar Supplier No. A13378.36
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 117-80-6 | C10H4Cl2O2 | 227.04 g/mol

Use in textiles, as organic catalyst, as chemical intermediate, as additive in dye binders. Also used as an important catalyst to produce 3,3-dichlorobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Use as seed disinfectant, textiles, as organic catalyst, as chemical intermediate, as additive in dye binders, as antifouling agent. Also used to control blue-green algae in ponds, lakes, and swimming pools. Also used as an important catalyst to produce 3,3-Dichloro benzine.

Solubility
Soluble in water (0.008g/L). Soluble in hot toluene almost transparency.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-80-6
Molecular Formula C10H4Cl2O2
Molecular Weight (g/mol) 227.04
MDL Number MFCD00001677
InChI Key SVPKNMBRVBMTLB-UHFFFAOYSA-N
Synonym 2,3-dichloro-1,4-naphthoquinone, dichlone, diclone, 2,3-dichloronaphthoquinone, phygon, algistat, sanquinon, uniroyal, phygon paste, dichloronaphthoquinone
PubChem CID 8342
SMILES ClC1=C(Cl)C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 193°C to 196°C (sublimation)
Boiling Point 275°C (2 mmHg)
Quantity 500 g
UN Number UN2811
Beilstein 1073511
Merck Index 14,3045
Solubility Information Soluble in water (0.008g/L). Soluble in hot toluene almost transparency.
IUPAC Name 2,3-dichloro-1,4-dihydronaphthalene-1,4-dione
Formula Weight 227.05
Percent Purity 98%
Chemical Name or Material 2,3-Dichloro-1,4-naphthoquinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.