missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%

Product Code. 10366270 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10366270 100 g Glass bottle 100g
10449020 10 g Glass bottle 10g
11915611 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10366270 Supplier Thermo Scientific Acros Supplier No. 113301000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 84-58-2 | C8Cl2N2O2 | 227.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-58-2
Molecular Formula C8Cl2N2O2
Molecular Weight (g/mol) 227.01
InChI Key HZNVUJQVZSTENZ-UHFFFAOYSA-N
Synonym 2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile
PubChem CID 6775
IUPAC Name 4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile
SMILES C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 210.0°C to 217.0°C
Color Yellow to Orange
Loss on Drying 0.5% max. (1 g, 80°C, 2 hrs, 40mm Hg vacuum)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Glass bottle
Quantity 100 g
Beilstein 10,902
Fieser 01,215; 02,112; 03,83; 04,130; 05,193; 06,168; 07,96; 08,153; 09,148; 10,135; 11,166; 13,104; 14,126; 15,125; 16,120
Merck Index 15,3072
Solubility Information Solubility in water: reacts. Other solubilities: soluble in benzene,dioxane,acetic acid,ethanol,slightly soluble in chloroform,methylenechloride
Formula Weight 227.01
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2,3-Dichloro-5,6-dicyano-1,4-benzoquinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.