missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-6-fluorobenzaldehyde, 97%, Thermo Scientific™

Product Code. 11406000
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406000 1 g 1g
11416000 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11406000 Supplier Thermo Scientific Alfa Aesar Supplier No. H26562.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as solvents. T2-Chloro-6-fluorobenzaldehyde was used in the synthesis of novel copolymers of methyl 2-cyano-3-dihalophenyl-2-propenoates and styrene

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95399-95-4
Molecular Formula C7H3Cl2FO
Molecular Weight (g/mol) 192.998
MDL Number MFCD04115870
InChI Key BDPIHKOPKWGJCU-UHFFFAOYSA-N
Synonym benzaldehyde, 2,3-dichloro-6-fluoro, 2,3-dichoro-6-fluorobenzaldehyde, 2-fluoro-5,6-dichlorobenzaldehyde
PubChem CID 3260062
IUPAC Name 2,3-dichloro-6-fluorobenzaldehyde
SMILES C1=CC(=C(C(=C1F)C=O)Cl)Cl

Specifications

Melting Point 56°C to 60°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 193.01
Percent Purity 97%
Chemical Name or Material 2,3-Dichloro-6-fluorobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.