missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzoic Acid, TRC

Product Code. 17931595
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17931595 10 g 10g
17901705 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17931595 Supplier Toronto Research Chemicals Supplier No. TRCD43195010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-45-3
Molecular Formula C7 H4 Cl2 O2
Molecular Weight (g/mol) 191.01
Synonym Benzoic acid, 2,3-dichloro-, 2,3-Dichlorobenzoic acid
IUPAC Name 2,3-dichlorobenzoic acid
SMILES OC(=O)c1cccc(Cl)c1Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H4Cl2O2/c8-5-3-1-2-4(6(5)9)7(10)11/h1-3H,(H,10,11)
Formula Weight 189.9588
Chemical Name or Material 2,3-Dichlorobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.