missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzylamine, 97%

Product Code. 11362687
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362687 1 g 1g
11372687 5 g 5g
11382687 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11362687 Supplier Thermo Scientific Alfa Aesar Supplier No. L11884.03
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 39226-95-4 | C7H7Cl2N | 176.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39226-95-4
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00047927
InChI Key JHBVZGONNIVXFJ-UHFFFAOYSA-N
Synonym 2,3-dichlorobenzylamine, 2,3-dichlorophenyl methanamine, 2,3-dichloro-benzylamine, 2,3-dichlorobenzyl amine, chembl13165, 1-2,3-dichlorophenyl methanamine, benzenemethanamine, 2,3-dichloro, 2,3-dichlorophenyl methylamine, pubchem16713, acmc-1ae0r
PubChem CID 587625
IUPAC Name (2,3-dichlorophenyl)methanamine
SMILES C1=CC(=C(C(=C1)Cl)Cl)CN

Specifications

Boiling Point 124°C to 126°C (15 mmHg)
Flash Point 105°C (221°F)
Quantity 1 g
UN Number UN2735
Beilstein 2639296
Formula Weight 176.05
Percent Purity 97%
Chemical Name or Material 2,3-Dichlorobenzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.