missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenol, 98+%

Product Code. p-7026750
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11410018

Brand: Thermo Scientific Alfa Aesar B22943.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 576-24-9 | C6H4Cl2O | 163.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 576-24-9
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002160
InChI Key UMPSXRYVXUPCOS-UHFFFAOYSA-N
Synonym phenol, 2,3-dichloro, dichlorophenol, unii-izm39u444l, ccris 5902, 2,3-dichloro phenol, 2,3 dichlorophenol, phenol,3-dichloro, pubchem3696, 2,3-dichloro-phenol, acmc-1ar0l
PubChem CID 11334
IUPAC Name 2,3-dichlorophenol
SMILES OC1=CC=CC(Cl)=C1Cl

Specifications

Melting Point 57°C to 60°C
Boiling Point 206°C
Flash Point 115°C (239°F)
Odor Phenol-like
Quantity 25 g
UN Number UN2020
Beilstein 2043615
Formula Weight 163
Percent Purity ≥98%
Chemical Name or Material 2,3-Dichlorophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.