missing translation for 'onlineSavingsMsg'
Learn More

2',3'-Dideoxyadenosine, 98%, Thermo Scientific Chemicals

Product Code. 15406469
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15406469 25 mg 25mg
15426469 50 mg 50mg
15416469 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15406469 Supplier Thermo Scientific Alfa Aesar Supplier No. J64573.MB
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4097-22-7 | Molecular Formula: C10H13N5O2 | Molecular Weight: 235.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4097-22-7
Molecular Formula C10H13N5O2
Molecular Weight (g/mol) 235.25
MDL Number MFCD00010534
InChI Key WVXRAFOPTSTNLL-NKWVEPMBSA-N
Synonym 2',3'-dideoxyadenosine, dideoxyadenosine, adenosine, 2',3'-dideoxy, ddado, 2s,5r-5-6-amino-9h-purin-9-yl tetrahydrofuran-2-yl methanol, ccris 428, unii-4q86ah641a, d2a, dsstox_cid_3771, dsstox_rid_77189
PubChem CID 20039
IUPAC Name [(2S,5R)-5-(6-aminopurin-9-yl)oxolan-2-yl]methanol
SMILES NC1=C2N=CN([C@H]3CC[C@@H](CO)O3)C2=NC=N1

Specifications

Melting Point 181°C to 184°C
Color White
Quantity 25 mg
Beilstein 619924
Merck Index 14,3101
Formula Weight 235.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2',3'-Dideoxyadenosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.