accessibility menu, dialog, popup

UserName

NH2NNNN(R)(S)OHO

CAS RN 4097-22-7

IUPAC Name: [(2S,5R)-5-(6-amino-9H-purin-9-yl)oxolan-2-yl]methanol
Synonyms: adenosine, 2',3'-dideoxy-
Molecular Weight (g/mol): 235.25
Molecular Formula: C10H13N5O2
InChi Key: WVXRAFOPTSTNLL-NKWVEPMBSA-N
SMILES: NC1=C2N=CN([C@H]3CC[C@@H](CO)O3)C2=NC=N1
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results