missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-4-hydroxybenzoic acid, 99%, Thermo Scientific Chemicals

Product Code. 11302049
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302049 1 g 1g
15453367 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302049 Supplier Thermo Scientific Alfa Aesar Supplier No. H26087.03
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175968-39-5
Molecular Formula C7H4F2O3
Molecular Weight (g/mol) 174.103
MDL Number MFCD01321116
InChI Key XIZIDHMVDRRFBT-UHFFFAOYSA-N
Synonym 4-carboxy-2,3-difluorophenol, 2,3-diifluoro-4-hydroxybenzoic acid, 2,3-difluoro-4-hydroxy-benzoic acid, rarechem al bo 1938, benzoic acid, 2,3-difluoro-4-hydroxy, pubchem2609, acmc-1c23d, 4-hydroxy-2,3-difluoro-benzoic acid, benzoic acid,2,3-difluoro-4-hydroxy
PubChem CID 2779307
IUPAC Name 2,3-difluoro-4-hydroxybenzoic acid
SMILES C1=CC(=C(C(=C1C(=O)O)F)F)O

Specifications

Melting Point 205°C to 210°C
Quantity 1 g
Formula Weight 174.1
Percent Purity 99%
Chemical Name or Material 2,3-Difluoro-4-hydroxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.