missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-4-hydroxybenzonitrile, 95%, Thermo Scientific Chemicals

Product Code. 15454927
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454927 1 g 1g
15464927 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454927 Supplier Thermo Scientific Alfa Aesar Supplier No. H32231.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126162-38-7
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.10
MDL Number MFCD00269596
InChI Key UIJJJWADIVZXNT-UHFFFAOYSA-N
PubChem CID 2736978
IUPAC Name 2,3-difluoro-4-hydroxybenzonitrile
SMILES OC1=C(F)C(F)=C(C=C1)C#N

Specifications

Melting Point 145°C to 149°C
Quantity 1 g
Synonym 4-cyano-2,3-difluorophenol, 2,3-difluoro-4-cyanophenol, 2,3-difluoro-4-cyano phenol, benzonitrile,2,3-difluoro-4-hydroxy, benzonitrile, 2,3-difluoro-4-hydroxy, 2,3-difluoro-4-hydroxybenzenecarbonitrile, pubchem2222, acmc-1c5t8, ksc514s9h, attercop-chm at134987
Formula Weight 155.1
Percent Purity 95%
Chemical Name or Material 2,3-Difluoro-4-hydroxybenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.