missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluorophenol, 98+%

Product Code. 11458806
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458806 1 g 1g
11468806 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458806 Supplier Thermo Scientific Alfa Aesar Supplier No. A16971.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6418-38-8 | C6H4F2O | 130.094 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6418-38-8
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.094
MDL Number MFCD00010262
InChI Key RPEPGIOVXBBUMJ-UHFFFAOYSA-N
Synonym phenol, 2,3-difluoro, phenol, difluoro, 2,3-difluoro phenol, difluorophenol, difluoro-phenol, 2.3-difluorophenol, 2,3-difluoro-phenol, pubchem1494, acmc-1b6pt, 2,3-bis fluoranyl phenol
PubChem CID 80879
IUPAC Name 2,3-difluorophenol
SMILES C1=CC(=C(C(=C1)F)F)O

Specifications

Melting Point 34°C to 38°C
Boiling Point 53°C to 55°C (25 mmHg)
Flash Point 56°C (132°F)
Odor Phenol-like
Quantity 1 g
UN Number UN2921
Beilstein 2082265
Formula Weight 130.09
Percent Purity ≥98%
Chemical Name or Material 2,3-Difluorophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.