missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydroxybenzoic acid, 99%

Product Code. 10081770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10081770 25 g Glass bottle 25g
10571591 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10081770 Supplier Thermo Scientific Acros Supplier No. 155170250
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 303-38-8 | C7H6O4 | 154.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303-38-8
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00002446
InChI Key GLDQAMYCGOIJDV-UHFFFAOYSA-N
Synonym pyrocatechuic acid, o-pyrocatechuic acid, 2-pyrocatechuic acid, 3-hydroxysalicylic acid, dobk, dhba, 2,3-dihydroxybenzoicacid, benzoic acid, 2,3-dihydroxy, catecholcarboxylic acid, 2,3-dihydroxy-benzoic acid
PubChem CID 19
ChEBI CHEBI:18026
IUPAC Name 2,3-dihydroxybenzoic acid
SMILES C1=CC(=C(C(=C1)O)O)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 204°C to 208°C
Color Beige to Brown or Gray-Pink
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (HO)2C6H3CO2H
Quantity 25 g
Beilstein 10, 375
Formula Weight 154.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Dihydroxybenzoic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Dihydroxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.