missing translation for 'onlineSavingsMsg'
Learn More

2',3-Dihydroxyflavone, 97%, Thermo Scientific Chemicals

Product Code. 11377429
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377429 1 g 1g
11387429 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377429 Supplier Thermo Scientific Alfa Aesar Supplier No. H27841.03
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6068-76-4 | Molecular Formula: C15H10O4 | Molecular Weight: 254.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6068-76-4
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.24
MDL Number MFCD00017674
InChI Key VECGDSZOFMYGAF-UHFFFAOYSA-N
Synonym 3,2'-dihydroxyflavone, 2',3-dihydroxyflavone, 3-hydroxy-2-2-hydroxyphenyl-4h-chromen-4-one, 2'-hydroxyflavonol, 3-hydroxy-2-2-hydroxyphenyl chromen-4-one, 4h-1-benzopyran-4-one, 3-hydroxy-2-2-hydroxyphenyl, 2'3-dioh-flavone, 3,2'-dihydroxy flavone, acmc-1b19t, 7,8-dihydroxyflavone-derivative-1
PubChem CID 455313
IUPAC Name 3-hydroxy-2-(2-hydroxyphenyl)chromen-4-one
SMILES OC1=CC=CC=C1C1=C(O)C(=O)C2=CC=CC=C2O1

Specifications

Melting Point 201°C to 204°C
Quantity 1 g
Formula Weight 254.24
Percent Purity 97%
Chemical Name or Material 2',3-Dihydroxyflavone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.