missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimercapto-1-propanol, 97.0-100.5%

Product Code. 10722184 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 10722184

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59-52-9 | C3H8OS2 | 124.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-52-9
Molecular Formula C3H8OS2
Molecular Weight (g/mol) 124.22
MDL Number MFCD00004864
InChI Key WQABCVAJNWAXTE-UHFFFAOYSA-N
Synonym dimercaprol, 2,3-dimercapto-1-propanol, 2,3-dimercaptopropanol, dithioglycerine, dicaptol, sulfactin, dimercaptopropanol, british anti-lewisite, 2,3-dithiopropanol, antoxol
PubChem CID 3080
ChEBI CHEBI:64198
IUPAC Name 2,3-bis(sulfanyl)propan-1-ol
SMILES C(C(CS)S)O

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2430g/mL
Boiling Point 120.0°C (15.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.0-100.5%
Packaging Glass bottle
Linear Formula HSCH2CH(SH)CH2OH
Refractive Index 1.5700 to 1.5740
Quantity 25 mL
Merck Index 15,3231
Specific Gravity 1.243
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol and vegetable oils
Formula Weight 124.22
Percent Purity 97.0-100.5%
Physical Form Liquid
Chemical Name or Material 2,3-Dimercapto-1-propanol, 97.0-100.5%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.