missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxybenzaldehyde, MedChemExpress

Product Code. 16421929
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16421929 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16421929 Supplier Medchem Express Supplier No. HY41407
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

MedChemExpress 2,3-Dimethoxybenzaldehyde (o-Veratraldehyde) is a benzaldehyde analog, with high antifungal activity (MIC=2.5 mM) 2,3-Dimethoxybenzaldehyde (o-Veratraldehyde) could be used for the synthesis of berberine.

2,3-Dimethoxybenzaldehyde (o-Veratraldehyde) is a benzaldehyde analog, with high antifungal activity (MIC=2.5 mM) 2,3-Dimethoxybenzaldehyde (o-Veratraldehyde) could be used for the synthesis of berberine.

Specifica

Chemical Name or Material 2,3-Dimethoxybenzaldehyde
Physical Form Solid
CAS 86-51-1
Color Off-White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C9H10O3
For Use With (Application) COVID-19-immunoregulation
Synonym o-Veratraldehyde 5,6-Dimethoxybenzaldehyde
Solubility Information DMSO : 100 mg/mL (601.79 mM; Need ultrasonic)
SMILES COC1=C(C=O)C=CC=C1OC
Molecular Weight (g/mol) 166.17
Formula Weight 166.17
Percent Purity 99.54%
Grade Research
Vedi altri risultati Mostra meno risultati

Research purposes only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.