missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxybenzoic acid, 99%

Product Code. 10060760 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10060760 25 g Glass bottle 25g
10604962 5 g Glass bottle 5g
10026233 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10060760 Supplier Thermo Scientific Acros Supplier No. 115420250
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1521-38-6 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1521-38-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00002432
InChI Key FODBVCSYJKNBLO-UHFFFAOYSA-N
Synonym o-veratric acid, benzoic acid, 2,3-dimethoxy, benzoic acid, dimethoxy, 2,3-dimethoxy benzoic acid, dimethoxybenzoic acid,2,3, o-veratric acid 6ci,7ci,8ci, 2,3-dimethoxybenzoicacid, dimethoxybenzoic acid, pubchem4948, 2,3-dimethoxy-benzoicaci
PubChem CID 15204
IUPAC Name 2,3-dimethoxybenzoic acid
SMILES COC1=CC=CC(C(O)=O)=C1OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 121°C to 124°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CO2H
Quantity 25 g
Beilstein 10, I, 174
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol
Formula Weight 182.18
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2, 3-Dimethoxybenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Dimethoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.