missing translation for 'onlineSavingsMsg'
Learn More

2(3)-tert-Butyl-4-methoxyphenol, 96%

Product Code. 11405547
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405547 250 g 250g
11495537 50 g 50g
11485537 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405547 Supplier Thermo Scientific Alfa Aesar Supplier No. B23530.30
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 25013-16-5 | C11H16O2 | 180.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2(3)-tert-Butyl-4-methoxyphenol is used as an industrial antioxidant. It is used as a food additive, preservative in food packaging, animal feed, cosmetics, rubber, and petroleum products.

Solubility
Soluble in ethanol, methanol, chloroform, fats, and oils. Insoluble in water.

Notes
Store in the dark place. Store away from light, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25013-16-5
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.247
MDL Number MFCD01779059
InChI Key MRBKEAMVRSLQPH-UHFFFAOYSA-N
Synonym 3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy
PubChem CID 8456
ChEBI CHEBI:76358
IUPAC Name 2-tert-butyl-4-methoxyphenol
SMILES CC(C)(C)C1=C(C=CC(=C1)OC)O

Specifications

Melting Point 55°C to 65°C
Quantity 250 g
Sensitivity Light sensitive
Merck Index 14,1547
Solubility Information Soluble in ethanol,methanol,chloroform,fats,and oils. Insoluble in water.
Formula Weight 180.25
Percent Purity 96%
Chemical Name or Material 2(3)-tert-Butyl-4-methoxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.