missing translation for 'onlineSavingsMsg'
Learn More

2(3H)-Benzothiazolone, TRC

Product Code. 30133427
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133427 1 g 1g
17933915 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133427 Supplier Toronto Research Chemicals Supplier No. TRCB2066601G
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 934-34-9
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.19
Synonym 2-Benzothiazolinone (6CI,7CI,8CI), 1,3-Benzothiazol-2(3H)-one, 1,3-Benzothiazol-2-ol, 2,3-Dihydro-1,3-benzothiazol-2-one, 2-Benzothiazolol, 2-Benzothiazolone, 2-Hydroxybenzothiazole, 2-Oxobenzothiazole, Benzothiazolone, NSC 26422, NSC 33823, NSC 7706
IUPAC Name 1,3-benzothiazol-2-ol
SMILES Oc1nc2ccccc2s1

Specifications

Analyte or Component Names 2(3H)-Benzothiazolone
Shipping Condition Room Temperature
InChI Formula InChI=1S/C7H5NOS/c9-7-8-5-3-1-2-4-6(5)10-7/h1-4H,(H,8,9)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 1 g
Formula Weight 151.0092
Chemical Name or Material 2(3H)-Benzothiazolone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.