missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichloroaniline, Dr. Ehrenstorfer

Product Code. 30317572
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317572 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317572 Supplier Dr. Ehrenstorfer Supplier No. DREC17700200100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,5-Trichloroaniline
CAS 636-30-6
Analyte or Component Names 2,4,5-Trichloroaniline
Shipping Condition +20°C
Molecular Formula C6 H4 Cl3 N
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C6H4Cl3N/c7-3-1-5(9)6(10)2-4(3)8/h1-2H,10H2
SMILES Nc1cc(Cl)c(Cl)cc1Cl
IUPAC Name 2,4,5-trichloroaniline
Molecular Weight (g/mol) 196.46
Quantity 100 mg
Formula Weight 194.9409 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.