missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluoro-N'-hydroxybenzenecarboximidamide, 97%, Thermo Scientific™

Product Code. 10167042
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10167042 1 g 1g
10027094 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10167042 Supplier Thermo Scientific Maybridge Supplier No. MO01290DA
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-34-9
Molecular Formula C7H5F3N2O
Molecular Weight (g/mol) 190.125
MDL Number MFCD06200878
InChI Key JWJFTTGUXZRMMI-UHFFFAOYSA-N
Synonym 2,4,5-trifluoro-n-hydroxy-benzamidine, 2,4,5-trifluoro-n-hydroxybenzimidamide, hydroxyimino 2,4,5-trifluorophenyl methylamine, z-2,4,5-trifluoro-n'-hydroxybenzene-1-carboximidamide
PubChem CID 9582832
IUPAC Name 2,4,5-trifluoro-N'-hydroxybenzenecarboximidamide
SMILES C1=C(C(=CC(=C1F)F)F)C(=NO)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 190.12
Percent Purity 97%
Chemical Name or Material 2, 4, 5-Trifluoro-N'-hydroxybenzenecarboximidamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.