missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorophenol, 94%

Product Code. 15473217
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15473217 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15473217 Supplier Thermo Scientific Alfa Aesar Supplier No. B20395.03
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2268-16-8 | C6H3F3O | 148.084 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2268-16-8
Melting Point 39°C to 42°C
Flash Point 55°C (131°F)
Odor Phenol-like
Molecular Formula C6H3F3O
MDL Number MFCD00061215
Quantity 1 g
UN Number UN2921
Synonym phenol, 2,4,5-trifluoro, phenol,2,4,5-trifluoro, 2,4,5-tfifluorophenol, 2,4,5-trifluoro-phenol, 2,4,5-?trifluorophenol, acmc-1cne2, 2,4,5-trifluorophenol, 2,4,5-tris fluoranyl phenol, 2,4,5-trifluorophenol 1gr
InChI Key ODGMYCITQAIRCI-UHFFFAOYSA-N
SMILES C1=C(C(=CC(=C1F)F)F)O
IUPAC Name 2,4,5-trifluorophenol
Molecular Weight (g/mol) 148.084
PubChem CID 123153
Formula Weight 148.09
Percent Purity 94%
Chemical Name or Material 2,4,5-Trifluorophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.