missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorophenylacetic acid, 94%

Product Code. 11396589
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11396589 1 g 1g
11306599 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396589 Supplier Thermo Scientific Alfa Aesar Supplier No. H26190.03
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 209995-38-0 | C8H5F3O2 | 190.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 209995-38-0
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00082479
InChI Key YSQLGGQUQDTBSL-UHFFFAOYSA-N
Synonym 2,4,5-trifluorophenylacetic acid, 2-2,4,5-trifluorophenyl acetic acid, 2,4,5-trifluorophenyl acetic acid, 2,4,5-trifluoro-phenyl-acetic acid, 2,4,5-trifluorophenylaceticacid, 2,4,5-trifluoro phenyl acetic acid, benzeneacetic acid, 2,4,5-trifluoro, pubchem3526, pubchem7287
PubChem CID 2777950
IUPAC Name 2-(2,4,5-trifluorophenyl)acetic acid
SMILES OC(=O)CC1=CC(F)=C(F)C=C1F

Specifications

Melting Point 121°C to 124°C
Quantity 1 g
Formula Weight 190.12
Percent Purity 94%
Chemical Name or Material 2,4,5-Trifluorophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.