missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorophenylboronic acid, 97%, Thermo Scientific™

Product Code. 11925791 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11925791 5 g 5g
11935791 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11925791 Supplier Thermo Scientific Acros Supplier No. 393800050
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 247564-72-3
Molecular Formula C6H4BF3O2
Molecular Weight (g/mol) 175.90
MDL Number MFCD01863165
InChI Key KCHHKNCSISEAAE-UHFFFAOYSA-N
Synonym 2,4,5-trifluorophenyl boronic acid, 2,4,5-trifluorobenzeneboronic acid, boronic acid, b-2,4,5-trifluorophenyl, boronic acid, 2,4,5-trifluorophenyl, 2,4,5-trifluorophenylboronicacid, pubchem1787, acmc-1cd74
PubChem CID 2783133
IUPAC Name (2,4,5-trifluorophenyl)boronic acid
SMILES OB(O)C1=CC(F)=C(F)C=C1F

Specifications

Melting Point 118.0°C to 119.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula F3C6H2B(OH)2
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 175.9
Percent Purity 97%
Chemical Name or Material 2, 4, 5-Trifluorophenylboronic acid
compliance-icons
Product Identifier
  • 2,4,5-Trifluorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.