missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trimethylphenol, 99%

Product Code. 11375449
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375449 250 mg 250mg
11385449 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11375449 Supplier Thermo Scientific Alfa Aesar Supplier No. H50427.MD
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 496-78-6 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-78-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00020050
InChI Key VXSCPERJHPWROZ-UHFFFAOYSA-N
Synonym pseudocumenol, phenol, 2,4,5-trimethyl, 5-hydroxypseudocumene, 1-hydroxy-2,4,5-trimethylbenzene, unii-3pca9e425c, acmc-1bmse, 4-06-00-03247 beilstein handbook reference
PubChem CID 10335
IUPAC Name 2,4,5-trimethylphenol
SMILES CC1=CC(=C(C=C1C)O)C

Specifications

Melting Point 72°C
Boiling Point 232°C
Odor Phenol-like
Quantity 250 mg
UN Number UN2430
Formula Weight 136.2
Percent Purity 99%
Chemical Name or Material 2,4,5-Trimethylphenol
compliance-icons
Product Identifier
  • 2,4,5-Trimethylphenol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2,4,5-Trimethylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.