missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tri-tert-butylphenol, 97%

Product Code. 10419052 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
50 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
500g
50g
5g
This item is not returnable. View return policy

Product Code. 10419052

Brand: Thermo Scientific Acros 139330050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 732-26-3 | C18H30O | 262.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 732-26-3
Molecular Formula C18H30O
Molecular Weight (g/mol) 262.44
MDL Number MFCD00008821
InChI Key PFEFOYRSMXVNEL-UHFFFAOYSA-N
Synonym 2,4,6-tri-tert-butylphenol, voidox, alkofen b, 2,4,6-tris tert-butyl phenol, 2,4,6-tri-t-butylphenol, phenol, 2,4,6-tris 1,1-dimethylethyl, phenol, 2,4,6-tri-tert-butyl, 2,4,6-tris 1,1-dimethylethyl phenol, unii-99j2vic675, tris 1,1-dimethylethyl phenol
PubChem CID 12902
SMILES CC(C)(C)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 129°C to 132°C
Boiling Point 277°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula [(CH3)3C]3C6H2OH
Quantity 5 g
UN Number 3077
Beilstein 06, IV, 3539
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
IUPAC Name 2,4,6-tritert-butylphenol
Formula Weight 262.42
Percent Purity 97%
Chemical Name or Material 2, 4, 6-Tri-tert-butylphenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.