missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichlorophenol, 98%

Product Code. 11444100
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444100 100 g 100g
11439173 250 g 250g
11429173 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444100 Supplier Thermo Scientific Alfa Aesar Supplier No. A11681.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 88-06-2 | C6H3Cl3O | 197.439 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2,4,6-Trichlorophenol, antiseptic, defoliant, and glue preservative, chemical intermediate, anti-mildew treatment, solvent for polyester fiber.

Solubility
Soluble in water(0.8g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-06-2
Molecular Formula C6H3Cl3O
MDL Number MFCD00002172
InChI Key LINPIYWFGCPVIE-UHFFFAOYSA-N
PubChem CID 6914
ChEBI CHEBI:28755
IUPAC Name 2,4,6-trichlorophenol

Specifications

Melting Point 63°C to 68°C
Density 1.675
Boiling Point 245°C to 246°C
Flash Point 99°C
Assay Percent Range 98%
Odor Phenol-like
Quantity 100 g
UN Number UN2020
Beilstein 776729
Merck Index 14,9644
Synonym dowicide 2 s, phenachlor, phenol, 2,4,6-trichloro, omal, phenaclor, rcra waste number u231, dowcide 2s, 1,3,5-trichloro-2-hydroxybenzene, 2,4,6-trichlorfenol, unii-mhs8c5bauz
Solubility Information Soluble aq. soln.(0.8 g/L).
SMILES OC1=C(Cl)C=C(Cl)C=C1Cl
Molecular Weight (g/mol) 197.44
Formula Weight 197.45
Percent Purity 98%
Chemical Name or Material 2,4,6-Trichlorophenol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,4,6-Trichlorophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2,4,6-Trichlorophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.