missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorobenzylamine, 97%, Thermo Scientific™

Product Code. 11405910
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405910 1 g 1g
11415910 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11405910 Supplier Thermo Scientific Alfa Aesar Supplier No. H26513.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.

Solubility
Slightly Soluble in water (1.2 g/L) (25°C).

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214759-21-4
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.13
MDL Number MFCD00236317
InChI Key RCHOKTKXVKKNBC-UHFFFAOYSA-N
Synonym 2,4,6-trifluorophenyl methanamine, 2,4,6-trifluorobenzylamine, 2,4,6-trifluorobenzyl amine, 2,4,6-trifluorobenzyl amin, 1-2,4,6-trifluorophenyl methylamine, benzenemethanamine, 2,4,6-trifluoro, 1-2,4,6-trifluorophenyl methanamine, pubchem16416, trifluorophenyl methanamine, ksc201i9f
PubChem CID 2777046
IUPAC Name (2,4,6-trifluorophenyl)methanamine
SMILES NCC1=C(F)C=C(F)C=C1F

Specifications

Refractive Index 1.471
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Slightly Soluble in water (1.2g/L) (25°C).
Formula Weight 161.13
Percent Purity 97%
Chemical Name or Material 2,4,6-Trifluorobenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.