missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluoropyrimidine, 98%

Product Code. 11381567
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381567 1 g 1g
11391567 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381567 Supplier Thermo Scientific Alfa Aesar Supplier No. L04404.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 696-82-2 | C4HF3N2 | 134.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-82-2
Molecular Formula C4HF3N2
Molecular Weight (g/mol) 134.06
MDL Number MFCD00039705
InChI Key NTSYSQNAPGMSIH-UHFFFAOYSA-N
Synonym pyrimidine, 2,4,6-trifluoro, pubchem6891, 2,4,6-trifluoropyrimide, 2,4, 6-trifluoropyrimidine, pyrimidine,2,4,6-trifluoro, pyrimidine, 2,4,6-trifluoro-6ci,7ci,8ci,9ci
PubChem CID 69677
IUPAC Name 2,4,6-trifluoropyrimidine
SMILES FC1=CC(F)=NC(F)=N1

Specifications

Boiling Point 100°C to 102°C
Refractive Index 1.405
Quantity 1 g
UN Number UN3265
Beilstein 607190
Sensitivity Moisture sensitive
Formula Weight 134.06
Percent Purity 98%
Chemical Name or Material 2,4,6-Trifluoropyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.