missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzenesulfonyl chloride, 97%

Product Code. 10397800 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10397800 25 g Glass bottle 25g
10666352 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10397800 Supplier Thermo Scientific Acros Supplier No. 152790250
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6553-96-4 | C15H23ClO2S | 302.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6553-96-4
Molecular Formula C15H23ClO2S
Molecular Weight (g/mol) 302.85
MDL Number MFCD00007433
InChI Key JAPYIBBSTJFDAK-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylbenzenesulfonyl chloride, 2,4,6-triisopropylbenzene-1-sulfonyl chloride, trisyl chloride, 2,4,6-triisopropylbenzenesulphonyl chloride, benzenesulfonyl chloride, 2,4,6-tris 1-methylethyl, 2,4,6-triisopropylbenzenesulfonylchloride, 2,4,6-triisopropyl-benzenesulfonyl chloride, tpscl 2,4,6-triisopropyl benzenesulfonyl chloride, 2,4,6-triisopropylphenylsulfonyl chloride, 2,4,6-triisopropyl benzenesulfonyl chloride
PubChem CID 81042
IUPAC Name 2,4,6-tri(propan-2-yl)benzenesulfonyl chloride
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)Cl)C(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 93°C to 97°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula [(CH3)2CH]3C6H2SO2Cl
Quantity 25 g
Fieser 01,1228; 03,308; 06,622
Solubility Information Solubility in water: decomposes
Formula Weight 302.85
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 2, 4, 6-Triisopropylbenzenesulfonyl chloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4,6-Triisopropylbenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.