missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzenesulfonyl Hydrazide, TRC

Product Code. 30452786
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452786 10 mg 10mg
30456823 100 mg 100mg
30453459 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30452786 Supplier Toronto Research Chemicals Supplier No. TRCT88618310MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39085-59-1
Molecular Formula C15H26N2O2S
Molecular Weight (g/mol) 298.44
IUPAC Name 2,4,6-tri(propan-2-yl)benzenesulfonohydrazide
SMILES CC(C)c1cc(C(C)C)c(c(c1)C(C)C)S(=O)(=O)NN

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H26N2O2S/c1-9(2)12-7-13(10(3)4)15(20(18,19)17-16)14(8-12)11(5)6/h7-11,17H,16H2,1-6H3
Formula Weight 298.1715
Chemical Name or Material 2,4,6-Triisopropylbenzenesulfonyl Hydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.