missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxybenzoic acid, 98%

Product Code. 11381787
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381787 5 g 5g
11391787 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381787 Supplier Thermo Scientific Alfa Aesar Supplier No. L04997.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 570-02-5 | C10H12O5 | 212.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 570-02-5
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.20
MDL Number MFCD00016497
InChI Key JATAKEDDMQNPOQ-UHFFFAOYSA-N
Synonym benzoic acid, 2,4,6-trimethoxy, 2,4,6-trimethoxy-benzoic acid, pubchem13726, acmc-1awjz, ksc495i9p, 2,4,6-trimethoxy benzoic acid
PubChem CID 68441
IUPAC Name 2,4,6-trimethoxybenzoic acid
SMILES COC1=CC(OC)=C(C(O)=O)C(OC)=C1

Specifications

Melting Point ∽158°C (decomposition)
Quantity 5 g
Beilstein 1972294
Formula Weight 212.2
Percent Purity 98%
Chemical Name or Material 2,4,6-Trimethoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.