missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxybenzonitrile, 98%

Product Code. 11415557
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415557 5 g 5g
11405557 1 g 1g
11425557 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11415557 Supplier Thermo Scientific Alfa Aesar Supplier No. B23540.06
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2571-54-2 | C10H11NO3 | 193.202 g/mol

2,4,6-Trimethoxybenzonitrile is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethoxybenzonitrile is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2571-54-2
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
MDL Number MFCD00001787
InChI Key GBRHJUMDNWLSCT-UHFFFAOYSA-N
PubChem CID 75731
IUPAC Name 2,4,6-trimethoxybenzonitrile
SMILES COC1=CC(=C(C(=C1)OC)C#N)OC

Specifications

Melting Point 140°C to 143°C
Odor Characteristic
Quantity 5 g
Synonym benzonitrile, 2,4,6-trimethoxy, 2,4,6-trimethoxy-benzonitrile, 2,4,6-trimethoxybenzenecarbonitrile, acmc-20aoeu, 2,4,6-trimethoxy benzonitrile, benzonitrile,2,4,6-trimethoxy, 2,4,6-trimethoxybenzonitrile
Solubility Information Insoluble in water.
Formula Weight 193.2
Percent Purity 98%
Chemical Name or Material 2,4,6-Trimethoxybenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.