Learn More
2,4,6-Trimethoxybenzonitrile, 98%
Beskrivelse
2,4,6-Trimethoxybenzonitrile is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,4,6-Trimethoxybenzonitrile is used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Tekniske data
Tekniske data
| Melting Point | 140°C to 143°C |
| Odor | Characteristic |
| Quantity | 25 g |
| Synonym | benzonitrile, 2,4,6-trimethoxy, 2,4,6-trimethoxy-benzonitrile, 2,4,6-trimethoxybenzenecarbonitrile, acmc-20aoeu, 2,4,6-trimethoxy benzonitrile, benzonitrile,2,4,6-trimethoxy, 2,4,6-trimethoxybenzonitrile |
| Solubility Information | Insoluble in water. |
| Formula Weight | 193.2 |
| Percent Purity | 98% |
| Chemical Name or Material | 2,4,6-Trimethoxybenzonitrile |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.