missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxytoluene, 97%

Product Code. 11361527
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361527 5 g 5g
11371527 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361527 Supplier Thermo Scientific Alfa Aesar Supplier No. L02155.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 14107-97-2 | C10H14O3 | 182.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14107-97-2
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00008375
InChI Key TZPKFPYZCMHDHL-UHFFFAOYSA-N
PubChem CID 84201
IUPAC Name 1,3,5-trimethoxy-2-methylbenzene
SMILES CC1=C(C=C(C=C1OC)OC)OC

Specifications

Melting Point 27°C to 28°C
Boiling Point 100°C to 105°C (1 mmHg)
Refractive Index 1.531
Quantity 5 g
Beilstein 2048952
Synonym 2,4,6-trimethoxytoluene, benzene, 1,3,5-trimethoxy-2-methyl, acmc-1bvuf, toluene, 2,4,6-trimethoxy, 1,3,5-trimethoxy-2-methyl-benzene, #, benzene,1,3,5-trimethoxy-2-methyl
Formula Weight 182.22
Percent Purity 97%
Chemical Name or Material 2,4,6-Trimethoxytoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.