missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide, Thermo Scientific Chemicals

Product Code. 15450437
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450437 10 mg 10mg
15460437 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450437 Supplier Thermo Scientific Alfa Aesar Supplier No. J65391.MA
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 313981-55-4 | Molecular Formula: C16H16F3NO2S | Molecular Weight: 343.36 g/mol

An inactive analog of m-3M3FBS. Used as a negative control.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An inactive analog of m-3M3FBS
  • Used as a negative control
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313981-55-4
Molecular Formula C16H16F3NO2S
Molecular Weight (g/mol) 343.36
MDL Number MFCD00784362
InChI Key SKJJIFRWCCSXGL-UHFFFAOYSA-N
Synonym o-3m3fbs, 2,4,6-trimethyl-n-2-trifluoromethyl phenyl benzenesulfonamide, tocris-1942, benzenesulfonamide,2,4,6-trimethyl-n-2-trifluoromethyl phenyl
PubChem CID 770820
IUPAC Name 2,4,6-trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide
SMILES CC1=CC(C)=C(C(C)=C1)S(=O)(=O)NC1=CC=CC=C1C(F)(F)F

Specifications

Color White
Quantity 10 mg
Formula Weight 343.36
Physical Form Powder
Chemical Name or Material 2,4,6-Trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.