missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzene-1-thiol, 97%, Thermo Scientific™

Product Code. 10009211
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10009211 10 g 10g
10295054 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10009211 Supplier Thermo Scientific Maybridge Supplier No. BTB12350EA
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1541-10-2
Molecular Formula C9H12S
Molecular Weight (g/mol) 152.255
MDL Number MFCD00218475
InChI Key PKANQZUPJCMBAK-UHFFFAOYSA-N
Synonym 2,4,6-trimethylthiophenol, 2,4,6-trimethylbenzene-1-thiol, benzenethiol,2,4,6-trimethyl, 2-mercaptomesitylene, 2-sulphanylmesitylene, acmc-20aoem, 2,4,6-trimethyl-benzene-1-thiol
PubChem CID 284449
IUPAC Name 2,4,6-trimethylbenzenethiol
SMILES CC1=CC(=C(C(=C1)C)S)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Density 1.0240g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)3C6H2SH
Quantity 10 g
Formula Weight 152.26
Percent Purity 97%
Chemical Name or Material 2, 4, 6-Trimethylbenzene-1-thiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.