missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylphenol, 98%

Product Code. 11494090
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494090 25 g 25g
11437297 100 g 100g
11447297 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494090 Supplier Thermo Scientific Alfa Aesar Supplier No. A15041.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 527-60-6 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 527-60-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002235
InChI Key BPRYUXCVCCNUFE-UHFFFAOYSA-N
Synonym mesitol, phenol, 2,4,6-trimethyl, mesityl alcohol, 2-hydroxymesitylene, 2,4,6-trimetylofenol, 1,3,5-trimethylphenol, 2,4,6-trimethyl-phenol, 1-hydroxy-2,4,6-trimethylbenzene, 2,4,6-trimethylofenol, unii-fpz32614n6
PubChem CID 10698
IUPAC Name 2,4,6-trimethylphenol
SMILES CC1=CC(=C(C(=C1)C)O)C

Specifica

Melting Point 70°C to 74°C
Boiling Point 220°C to 221°C
Quantity 25 g
UN Number UN2430
Formula Weight 136.2
Percent Purity 98%
Chemical Name or Material 2,4,6-Trimethylphenol

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.