missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylpyrylium tetrafluoroborate, 98+%

Product Code. 11380054
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11380054 5 g 5g
11390054 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11380054 Supplier Thermo Scientific Alfa Aesar Supplier No. L00467.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 773-01-3 | C8H11BF4O | 209.98 g/mol

It is used in the radical Arylation Reactions of 4, 6, 8-Trimethylazulene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the radical Arylation Reactions of 4, 6, 8-Trimethylazulene.

Solubility
Soluble in water and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 773-01-3
Molecular Formula C8H11BF4O
Molecular Weight (g/mol) 209.98
MDL Number MFCD00032098
InChI Key PYOPRIFMELCENN-UHFFFAOYSA-N
Synonym 2,4,6-trimethylpyrylium tetrafluoroborate, 2,4,6-trimethyl-1??-pyran-1-ylium tetrafluoroborate, 2,4,6-trimethylpyrylium tetrafluoborate, 2,4,6-trimethylpyryliumtetrafluoroborate, pyrylium, 2,4,6-trimethyl-, tetrafluoroborate 1-, pyrylium, 2,4,6-trimethyl-, tetrafluoroborate 1-1:1, 2,4,6-trimethyl-1$l^ 4-pyran-1-ylium tetrafluoroborate
PubChem CID 164732
IUPAC Name 2,4,6-trimethylpyrylium;tetrafluoroborate
SMILES F[B-](F)(F)F.CC1=CC(C)=[O+]C(C)=C1

Specifications

Melting Point 224°C to 226°C
Quantity 5 g
Beilstein 3639361
Solubility Information Soluble in water and methanol.
Formula Weight 209.98
Percent Purity ≥98%
Chemical Name or Material 2,4,6-Trimethylpyrylium tetrafluoroborate
compliance-icons
Product Identifier
  • 2,4,6-Trimethylpyrylium tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2,4,6-Trimethylpyrylium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.