missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(trifluoromethyl)-1,3,5-triazine, 98%

Product Code. 11385035
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11385035 1 g 1g
11499477 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11385035 Supplier Thermo Scientific Alfa Aesar Supplier No. B22001.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 368-66-1 | C6F9N3 | 285.07 g/mol

2,4,6-Tris(trifluoromethyl)-1,3,5-triazine is used to prepare 1-phenyl-4,6-bis-trifluoromethyl-1H-pyrazolo[3,4-d]pyrimidine by reacting with sodium 5-amino-1-phenyl-1H-pyrazole-4-carboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Tris(trifluoromethyl)-1,3,5-triazine is used to prepare 1-phenyl-4,6-bis-trifluoromethyl-1H-pyrazolo[3,4-d]pyrimidine by reacting with sodium 5-amino-1-phenyl-1H-pyrazole-4-carboxylate.

Notes
Air and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368-66-1
Molecular Formula C6F9N3
Molecular Weight (g/mol) 285.07
MDL Number MFCD00042436
InChI Key LSGBKABSSSIRJF-UHFFFAOYSA-N
Synonym 2,4,6-tris trifluoromethyl-1,3,5-triazine, s-triazine, 2,4,6-tris trifluoromethyl, 1,3,5-triazine, 2,4,6-tris trifluoromethyl, 2,4,6-tris trifluoromethyl-sym-triazine, 2,4,6-tris trifluoromethyl-s-triazine, tris trifluoromethyl-1,3,5-triazine, acmc-1cp6j, 5-26-01-00320 beilstein handbook reference, 1,3,5-triazine,2,4,6-tris trifluoromethyl, 2,4,6-tris trifluoromethyl-1,3,5 triazine
PubChem CID 9715
IUPAC Name 2,4,6-tris(trifluoromethyl)-1,3,5-triazine
SMILES FC(F)(F)C1=NC(=NC(=N1)C(F)(F)F)C(F)(F)F

Specifications

Density 1.596
Boiling Point 95°C to 96°C
Flash Point None
Refractive Index 1.322
Quantity 1 g
Beilstein 302339
Formula Weight 285.07
Percent Purity 98%
Chemical Name or Material 2,4,6-Tris(trifluoromethyl)-1,3,5-triazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.