missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Benzyloxyphenyl)ethanol, 98+%

Product Code. 11374086
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374086 25 g 25g
11364086 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374086 Supplier Thermo Scientific Alfa Aesar Supplier No. L19217.14
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 61439-59-6 | C15H16O2 | 228.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61439-59-6
Molecular Formula C15H16O2
Molecular Weight (g/mol) 228.29
MDL Number MFCD00017532
InChI Key JCUJAHLWCDISCC-UHFFFAOYSA-N
Synonym 2-4-benzyloxyphenyl ethanol, 2-4-benzyloxy phenyl ethanol, p-benzyloxy phenethyl alcohol, 4-benzyloxyphenylethyl alcohol, 4-benzyloxyphenethyl alcohol, 4-benzyloxy phenethylalcohol, benzeneethanol, 4-phenylmethoxy, 2-p-benzyloxyphenyl ethanol, 4-benzyloxy phenethyl alcohol
PubChem CID 3017260
IUPAC Name 2-(4-phenylmethoxyphenyl)ethanol
SMILES OCCC1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 85°C to 87°C
Quantity 25 g
Beilstein 2054624
Formula Weight 228.29
Percent Purity ≥98%
Chemical Name or Material 2-(4-Benzyloxyphenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.