missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Product Code. 11435596
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435596 5 g 5g
11425596 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11435596 Supplier Thermo Scientific Alfa Aesar Supplier No. B20731.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32890-87-2
Molecular Formula C9H4F6O2
Molecular Weight (g/mol) 258.119
MDL Number MFCD00042496
InChI Key JTOIZLCQNWWDCN-UHFFFAOYSA-N
Synonym 2,4-bis trifluoromethyl benzoic acid, 2,4-bis-trifluoromethylbenzoic acid, 2,4-di trifluoromethyl benzoic acid, benzoic acid, 2,4-bis trifluoromethyl, 2,4-bis-trifluoromethyl-benzoic acid, maybridge4_003283, acmc-20aoeb, pubchem8491, rarechem al bo 0512
PubChem CID 141752
IUPAC Name 2,4-bis(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)O

Specifications

Melting Point 108°C to 110°C
Quantity 5 g
Beilstein 2057460
Formula Weight 258.12
Percent Purity 98%
Chemical Name or Material 2,4-Bis(trifluoromethyl)benzoic acid
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.