missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis(trifluoromethyl)benzyl bromide, 95%

Product Code. 10812621 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
10812621 1 g 1g
10301760 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 10812621

Brand: Thermo Scientific Acros 436350010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 140690-56-8 | C9H5BrF6 | 307.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140690-56-8
Molecular Formula C9H5BrF6
Molecular Weight (g/mol) 307.03
MDL Number MFCD00010306
InChI Key SWFFFUJOWAJJCH-UHFFFAOYSA-N
Synonym 2,4-bis trifluoromethyl benzyl bromide, 1-bromomethyl-2,4-bis trifluoromethyl benzene, 2,4-bis trifluoromethyl benzylbromide, benzene, 1-bromomethyl-2,4-bis trifluoromethyl, 2,4-bis trifluoromethyl-1-bromomethyl benzene, pubchem4879, acmc-1c0tm, 2,4-bis-trifluoromethyl-benzylbromide, 2,4-bis-trifluoromethyl benzyl bromide, 2,4-bis-trifluoromethyl-benzyl bromide
PubChem CID 518871
IUPAC Name 1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)CBr

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 307.03
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2, 4-Bis(trifluoromethyl)benzyl bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Bis(trifluoromethyl)benzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.