missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(Bromomethyl)phenyl]propionic acid, 96%

Product Code. 15457258
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457258 25 g 25g
15447258 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457258 Supplier Thermo Scientific Alfa Aesar Supplier No. H60763.14
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111128-12-2 | C10H11BrO2 | 243.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111128-12-2
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD02093445
InChI Key QQXBRVQJMKBAOZ-UHFFFAOYSA-N
Synonym 2-4-bromomethyl phenyl propanoic acid, 2-4-bromomethyl phenyl propionic acid, 2-4-bromomethyl phenylpropionic acid, 2-4-bromomethyl-phenyl-propionic acid, 4-bromomethyl hydratropic acid, 2-4-bromomethyl phenyl propanic acid, 2-p-bromomethyl phenyl propionic acid, 2-4'-bromomethylphenyl propionic acid, 2-4-bromomethyl phenylproplonic acid
PubChem CID 2733978
IUPAC Name 2-[4-(bromomethyl)phenyl]propanoic acid
SMILES CC(C1=CC=C(C=C1)CBr)C(=O)O

Specifications

Melting Point 126°C to 130°C
Quantity 25 g
UN Number UN3261
Formula Weight 243.1
Percent Purity 96%
Chemical Name or Material 2-[4-(Bromomethyl)phenyl]propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.