missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenoxy)-6-methylpyrazine, 97%, Thermo Scientific™

Product Code. 10163764
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10163764 1 g Amber Glass Bottle 1g
10798835 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10163764 Supplier Thermo Scientific Maybridge Supplier No. CC62710DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 915707-62-9
Molecular Formula C11H9BrN2O
Molecular Weight (g/mol) 265.11
MDL Number MFCD09702408
InChI Key VJZHVPRWFQZLGM-UHFFFAOYSA-N
Synonym 2-4-bromophenoxy-6-methylpyrazine, pyrazine, 2-4-bromophenoxy-6-methyl, 4-bromo-1-6-methylpyrazin-2-yloxy benzene
PubChem CID 24229692
IUPAC Name 2-(4-bromophenoxy)-6-methylpyrazine
SMILES CC1=CN=CC(=N1)OC2=CC=C(C=C2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 265.11
Percent Purity 97%
Chemical Name or Material 2-(4-Bromophenoxy)-6-methylpyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.