missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)furan, ≥97%, Thermo Scientific™

Product Code. 10709595
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10709595 250 mg 250mg
10431134 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10709595 Supplier Thermo Scientific Maybridge Supplier No. CC34410CB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 14297-34-8
Molecular Formula C10H7BrO
Molecular Weight (g/mol) 223.069
MDL Number MFCD07775614
InChI Key FBIKSVALROQWIY-UHFFFAOYSA-N
Synonym 2-4-bromophenyl furan, 2-4-bromophenyl-furan, pubchem2039, 2 4-bromophenyl furan, 2-p-bromophenyl furan, 4-bromo-2-furyl benzene, furan, 2-4-bromophenyl
PubChem CID 11276114
IUPAC Name 2-(4-bromophenyl)furan
SMILES C1=COC(=C1)C2=CC=C(C=C2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 223.07
Percent Purity 97%
Chemical Name or Material 2-(4-Bromophenyl)furan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.