missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chloro-2-fluorophenyl)acetonitrile, 97%, Thermo Scientific™

Product Code. 10049391
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10049391 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10049391 Supplier Thermo Scientific Maybridge Supplier No. KM10105DA
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 75279-53-7
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H5ClFN
MDL Number MFCD01312758
Quantity 1 g
Synonym 4-chloro-2-fluorophenylacetonitrile, 2-4-chloro-2-fluorophenyl acetonitrile, 2-fluoro-4-chlorobenzyl cyanide, 4-chloro-2-fluorobenzyl cyanide, benzeneacetonitrile, 4-chloro-2-fluoro, 4-chloro-2-fluoro-phenyl-acetonitrile, 4-chloro-2-fluoro-phenyl acetonitrile, 2-4-chloro-2-fluorophenyl ethanenitrile, pubchem4296, ksc494e1b
InChI Key RTQOANCZEAIDEZ-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Cl)F)CC#N
IUPAC Name 2-(4-chloro-2-fluorophenyl)acetonitrile
Molecular Weight (g/mol) 169.583
PubChem CID 2736569
Formula Weight 169.59
Percent Purity 97%
Chemical Name or Material 2-(4-Chloro-2-fluorophenyl)acetonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.