missing translation for 'onlineSavingsMsg'
Learn More

2-[4-(Chloromethyl)phenyl]-1,3-thiazole, 97%, Thermo Scientific™

Product Code. 10396804
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10396804 250 mg Amber Glass Bottle 250mg
10783964 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10396804 Supplier Thermo Scientific Maybridge Supplier No. CC40224CB
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-61-2
Molecular Formula C10H8ClNS
Molecular Weight (g/mol) 209.691
MDL Number MFCD09064951
InChI Key BFAWMDHRKPPXSO-UHFFFAOYSA-N
Synonym 2-4-chloromethyl phenyl-1,3-thiazole, 2-4-chloromethyl phenyl thiazole
PubChem CID 24229488
IUPAC Name 2-[4-(chloromethyl)phenyl]-1,3-thiazole
SMILES C1=CC(=CC=C1CCl)C2=NC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 209.7
Percent Purity 97%
Chemical Name or Material 2-[4-(Chloromethyl)phenyl]-1, 3-thiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.