missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenoxy)-2-methylpropionic acid, 99%, Thermo Scientific Chemicals

Product Code. 10572591 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10572591 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10572591 Supplier Thermo Scientific Acros Supplier No. 189092500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 882-09-7
Molecular Formula C10H11ClO3
Molecular Weight (g/mol) 214.65
MDL Number MFCD00004192
InChI Key TXCGAZHTZHNUAI-UHFFFAOYSA-N
Synonym clofibric acid, 2-4-chlorophenoxy-2-methylpropanoic acid, chlorofibrinic acid, clofibrinic acid, chlorfibrinic acid, clofibrin, chlorophibrinic acid, pcib, 2-4-chlorophenoxy-2-methylpropionic acid, clofibrate free acid
PubChem CID 2797
ChEBI CHEBI:34648
IUPAC Name 2-(4-chlorophenoxy)-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)OC1=CC=C(C=C1)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 120.0°C to 123.0°C
Color Brown to Yellow
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula ClC6H4OC(CH3)2CO2H
Quantity 250 g
Merck Index 15, 2371
Formula Weight 214.65
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-(4-Chlorophenoxy)-2-methylpropionic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.