missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenoxy)isobutyric acid, 98%, Thermo Scientific™

Product Code. 11458257
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458257 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11458257 Supplier Thermo Scientific Alfa Aesar Supplier No. A17624.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 882-09-7
Molecular Formula C10H11ClO3
Molecular Weight (g/mol) 214.645
MDL Number MFCD00004192
InChI Key TXCGAZHTZHNUAI-UHFFFAOYSA-N
Synonym clofibric acid, 2-4-chlorophenoxy-2-methylpropanoic acid, chlorofibrinic acid, clofibrinic acid, chlorfibrinic acid, clofibrin, chlorophibrinic acid, pcib, 2-4-chlorophenoxy-2-methylpropionic acid, clofibrate free acid
PubChem CID 2797
ChEBI CHEBI:34648
IUPAC Name 2-(4-chlorophenoxy)-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)OC1=CC=C(C=C1)Cl

Specifications

Melting Point 119°C to 120°C
Quantity 25 g
Beilstein 1874067
Merck Index 14,2378
Formula Weight 214.65
Percent Purity 98%
Chemical Name or Material 2-(4-Chlorophenoxy)isobutyric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.